Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bump caps

Bump caps

Bump cap, regular fit, PW59, Dark blue
Catalog # PRTWPW59NAR
Supplier:   Portwest
Bump caps
Catalog # PRTWPW59NAR
Supplier:  Portwest
Bump caps
Bump caps

Bump cap, regular fit, PW59, Dark blue

Price
18,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

These classic design bump caps features ventilation ports for maximum breathability and metallic ring holes for baseball cap look.

  • Inner ABS shell and soft pad for shock absorption
  • Retail bag which aids presentation for retail sales
  • Hook and loop adjustable size
  • Made from 100% cotton

Long peak (7 cm) for maximum protection against shock and light. The durable high-density EVA protects and provides comfort to the wearer.

Certifications: EN 812

Packaging: 60 caps per carton.

Documents

Specifications
  • Colour:
    Dark blue
  • Description:
    Bump cap, regular fit
  • Model:
    PW59
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.