ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP) ≥99.5% (by GC), LiChrosolv® for liquid chromatography, Supelco®
Health
Irritant
N-Methyl-2-pyrrolidone (NMP) ≥99.5% (by GC), LiChrosolv® for liquid chromatography, Supelco®
Supplier:  Merck
Certificates
undefined
Health
Irritant
N-Methyl-2-pyrrolidone (NMP) ≥99.5% (by GC), LiChrosolv® for liquid chromatography, Supelco®
Supplier:  Merck
Certificates
Specification Test Results
  • Purity (GC):
    ≥ 99.5 %
  • Water:
    ≤ 0.05 %
  • Transmission (at 300 nm):
    ≥ 50 %
  • Transmission (at 325 nm):
    ≥ 90 %
  • Transmission (at 350 nm):
    ≥ 97 %
  • Transmission (at 400 nm):
    ≥ 99 %