Screw cap with septum for sample tubes
Catalog # SIMPT504AQX
Fournisseur: Simport Scientific
Spécifications
- Description:Septum screw cap for sample tubes
- Material:PP
- Pk:250
Spécifications
À propos de cet article
This aqua colour screw cap incorporates a pierceable septum made of chemically resistant PTFE on the outside and silicone on the inside, both components being stable over a broad range of temperatures.
- Acts as a silicone O-ring for better sample protection
- Especially made to fit and be used with all Simport® sample tubes
The cap is pierceable with pipette tips as well as with syringe needles.