ContactUsLinkComponent
Head tops for respiratory system Versaflo™, M-206 series
Head tops for respiratory system Versaflo™, M-206 series
Catalog # 111-2402
Supplier:  3M
undefined
Head tops for respiratory system Versaflo™, M-206 series
Catalog # 111-2402
Supplier:  3M
Specifications
  • Description:
    Faceshield M-206, with comfort faceseal with clear visor, for dusts, spraying and chemical processing
  • Pk:
    1
Specifications
About this item
Lightweight, compact and well-balanced faceshield with a comfortable faceseal for dusts, spraying and chemical processing.
  • Integrated protection from a range of respiratory, face, bumpcap, eye and hearing hazards
  • Increased control and comfort with the deflector which allows users to direct the airflow inside the headtop
  • Easy to maintain: Spare parts and accessories are quick, easy and intuitive to replace
  • Visor design combines excellent peripheral and downward vision with good optical clarity
  • Coated PC visor provides added chemical and scratch resistance
Recommended industries: Automotive, construction, metal production and fabrication, transportation.
The M-206 faceshield offers respiratory, bumpcap, eye and face (EN 166 Medium Impact) protection. The helmet features a comfortable general purpose faceseal for dusts, spraying and chemical processing. The M-206 helmet meets the performance requirements (TH3) for this type of product when used with specific 3M™ Versaflo™ Air Delivery Units.
Protection level:
NPF: 200 (V-Series), NPF: 50 (Jupiter), NPF: 500 (TR-302E/TR-602E)