Order Entry
Belgium
ContactUsLinkComponent
 
Qscript lyo 1-step
Catalog # 733-2926
Supplier:  Quantabio
CAS Number:  
Qscript lyo 1-step
Catalog # 733-2926
Supplier:  Quantabio
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Application:
    RT-qPCR probe
  • Multiplexing:
    Yes
  • One step:
    Yes
  • Description:
    Qscript lyo 1-step, 24 reactions
  • Seq:
    02

Specifications

About this item

Qscript lyo 1-step is a lyophilised single-reaction reagent optimised for highly sensitive and reproducible one-step RT-qPCR using hydrolysis probes.

  • Lyophilised single tube format – easy to use, reduces cross-contamination
  • High sensitivity and specificity – detect as low as 0,5 pg RNA
  • Wide dynamic range from 0,5 to 500 pg RNA
  • Superior multiplexing with up to five targets per reaction
  • Eliminate freezer storage with up to nine months stability at room temperature

Qscript lyo 1-step reagent contains a hot-start thermo-stable polymerase, and a genetically engineered reverse transcriptase, as well as other components to ensure higher performance detection of up to 5 targets with maximum sensitivity and specificity. The enhanced stability of the freeze-dried master mix enables convenient shipping and storage at room temperature. The single tube reaction facilitates easy reaction set up while preventing potential cross-contamination.