Order Entry
Belgium
ContactUsLinkComponent
Sputofluol® for microbiology, for microscopy, Sigma-Aldrich®
Corrosive
Environment
Sputofluol® for microbiology, for microscopy, Sigma-Aldrich®
Catalog # 1.08000.1000
Supplier:  Merck
CAS Number:  
undefined
Corrosive
Environment
Sputofluol® for microbiology, for microscopy, Sigma-Aldrich®
Catalog # 1.08000.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Pk Info:
    Preparation Capacity 1000 applications

Specifications

Specification Test Results

  • NaOH content (acidimetric):
    7 - 8%
  • Assay (iodometric, free Cl₂):
    ≥4,5%

Specification Test Results

About this item

 :
ADR:8,II
 :M-Clarity™ quality level = MQ300
UN:1791
 :GHS05
 :GHS09
Storage Temperature:Ambient
 :1