ContactUsLinkComponent
Heavy duty hoses for airhoods
Heavy duty hoses for airhoods
Catalog # NORHA161253
Supplier:  Honeywell Safety
Heavy duty hoses for airhoods
Catalog # NORHA161253
Supplier:  Honeywell Safety
Specifications
  • Colour:
    Black
  • Length:
    10 m
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Black heavy duty breathing hoses for use in connection with light and heavy duty airhoods.
  • Anti-static
  • With quick connection double action safety male and female couplings
  • Available in several lengths
Certifications: EN 139