Order Entry
Belgium
ContactUsLinkComponent
Storage box, PCR, with Lid
Storage box, PCR, with Lid
Catalog # NEOL2-1914
Supplier:  NEOLAB
Storage box, PCR, with Lid
Catalog # NEOL2-1914
Supplier:  NEOLAB

Specifications

  • Colour:
    Blue
  • Description:
    PCR storage box, 133×133×30 mm
  • Material:
    PP
  • Pk:
    1

Specifications

About this item

The PCR storage box is suitable for 0,2 ml PCR tubes and strips.

  • Stackable
  • Alphanumerical coding on the bottom

Features grid partition of 14×14, suitable for 196 single tubes and 12-tube strips with lids.

It has an extra flat lid which prevents the PCR tubes and strips from falling out.