Order Entry
Belgium
ContactUsLinkComponent
Propidium iodide solution 1.0 mg/ml in water
Propidium iodide solution 1.0 mg/ml in water
Catalog # P4864-10ML
CAS Number:  
undefined
Propidium iodide solution 1.0 mg/ml in water
Catalog # P4864-10ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance (Color):
    Faint Orange to Very Dark Orange and Faint Red to Very Dark Red and Faint Purple to Very Dark Purple and Orange-Red and Red-Orange
  • Appearance (Turbidity):
    Clear
  • Appearance (Form), 1.0 mg/mL, H₂O (Solution):
    Liquid
  • Purity (HPLC):
    ≥94%
  • Extinction Coefficient, 491 to 495 nm in phosphate buffer, pH 7:
    5400 - 6400
  • Wavelength :
    491 - 495 nm

Specification Test Results

About this item

 :
 :668.4 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
CAS Number:25535-16-4
Formula:C₂₇H₃₄I₂N₄
MDL Number:MFCD00011921