Orders LinkContactUsLinkComponent
Dropping funnels, cylindrical, with double jacket
Dropping funnels, cylindrical, with double jacket
Dropping funnels, cylindrical, with double jacket
Borosilicate glass 3.3, with PTFE tap.
  • Graduated
  • With drop counter
  • with GL 14 olives (PE)
  • With pressure compensation
Product Family Options
  • Description
    Capacity
    Socket size
  • Dropping funnel with drilling 2,5 mm
    50 ml
    14/23 NS
    1 * 1 ST
    1 * 1 ST - 585,00 €
    585,00 €
    1 * 1 ST
       585,00 €
  • Dropping funnel with drilling 2,5 mm
    100 ml
    19/26 NS
    1 * 1 ST
    1 * 1 ST - 633,00 €
    633,00 €
    1 * 1 ST
       633,00 €