Order Entry
Belgium
ContactUsLinkComponent
 
Acetate standard for ion chromatography, TraceCERT®, Supelco®
Catalog # 51791-100ML
Supplier:  Merck
CAS Number:  
Acetate standard for ion chromatography, TraceCERT®, Supelco®
Catalog # 51791-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Matrix:
    H₂O
  • Pk:
    100 mL
  • Description:
    Acetate standard solution

Specifications

About this item

Prepared with acetic acid, NaOH and water (pH 7 to 8); filtered through a 0,2 μm membrane filter and stabilised with sodium azide.

Certifications: Certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034.