Order Entry
Belgium
Orders LinkContactUsLinkComponent
Hemoglobin standard

Hemoglobin standard

Hemoglobin standard, 500 µM heme, 1 ml
Catalog # CAYM700543-1
Supplier:   Cayman Chemical
Hemoglobin standard
Catalog # CAYM700543-1
Supplier:  Cayman Chemical
CAS Number:  
Hemoglobin standard

Hemoglobin standard, 500 µM heme, 1 ml

CAS Number:  
Price
46,50 €
Log in for availability
1 * 1 mL
Product Details & Documents

Ready for use with colorimetric assay kit. This vial contains solution of 500 µM heme.

Stability: ≥1 year at 4 °C.

Caution: For laboratory use only. Not for human or veterinary use.

Specifications

Specifications

  • Pk:
    1 ml
  • Description:
    Hemoglobin standard
  • Pk Info:
    500 µM heme

Specifications