Ethanol absolute (untaxed) ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
:64-17-5
:
:C₂H₅OH
:MFCD00003568
:46.07 g/mol
:78…79 °C (1.013 hPa)
:–115 °C
:0.790…0.791 g/cm3 (20 °C)
:12 °C
:Ambient
:200-578-6
:1170
:3,II
:GHS07
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Assay (GC):Min. 99.8 Vol.%
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.2 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Transmittance at 210 nm:Min. 20 %
- Transmittance at 225 nm:Min. 50 %
- Transmittance at 240 nm:Min. 80 %
- Transmittance from 260 nm:Min. 98 %
- Fluorescence (chinin) at 254 nm:Max. 1 ppb
- Fluorescence (chinin) at 365 nm:Max. 1 ppb
- Autoignition Temperature:683 °F
- Vapor Density:1.59 (vs air)
Frequently Bought Together