Orders LinkContactUsLinkComponent
Swab sampling kit
Swab sampling kit
Swab sampling kit, incl. 10 ml buffer (in tube) and swab with PS shaft and polyester head
Catalog # TSCLTS/5/42
Supplier:   TSC LIMITED
Swab sampling kit
Catalog # TSCLTS/5/42
Supplier:  TSC LIMITED
Swab sampling kit
Swab sampling kit
Swab sampling kit, incl. 10 ml buffer (in tube) and swab with PS shaft and polyester head
Price
151,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
This kit includes 10 ml neutralising buffer in a tube and a sampling swab with PS (breakable) shaft and polyester (dacron) tip, packed in a peel pouch.
  • Gamma irradiated (Sterility Assurance Level (SAL) of 10⁻⁶)
  • Accurate volumes for serial dilution techniques
Recommended for hygiene and environmental monitoring of equipment or surfaces testing where traces of disinfectants are possibly present. Ideal for maintaining and transporting microbiological samples & eliminating any traces of sanistisers or disinfectants remaining on surfaces.
Excellent recovery of organisms including: TVC, Listeria spp., Staphylococci, Enterobacteriaceae eg Salmonella, E.coli (E coli / ecoli / e. coli), Enterobacter, staphylococcus, klebsiella, E. coli Enterobacter, Enterobacter sakazakii.
Specifications
  • Description:
    Swab sampling kit, incl. 10 ml buffer (in tube) and swab with PS shaft and polyester head