Specific Gravity Hydrometers
Supplier: Amarell
About this item
These general-purpose hydrometers are available with a thermometer inside the body or without a thermometer.
- Capillary prismatic
- Reliable and simple
- Paper scale with type designation
- Shot weighted body
: Supplied in a transparent plastic case.
53 Options Available Below
Product Details & Documents
These general-purpose hydrometers are available with a thermometer inside the body or without a thermometer.
- Capillary prismatic
- Reliable and simple
- Paper scale with type designation
- Shot weighted body
: Supplied in a transparent plastic case.
Specifications for Product Family
Specifications
- Catalog No:
- AMARH826760-FL
- AMARH826764-FL
- AMARH826780
- AMARH826782
- AMARH826784
- AMARH826786
- AMARH826788
- AMARH826790
- AMARH826792
- AMARH826910
- AMARH826912
- AMARH826914
- AMARH826916
- AMARH826918
- AMARH826920
- AMARH826922
- AMARH826924
- AMARH826926
- AMARH826928
- AMARH826930
- AMARH826932
- AMARH826934
- AMARH826936
- AMARH826938
- AMARH826940
- AMARH826942
- AMARH826500
- AMARH826502
- AMARH826504
- AMARH826506
- AMARH826508
- AMARH826510
- AMARH826512
- AMARH826628
- AMARH826514
- AMARH826630
- AMARH826516
- AMARH826632
- AMARH826634
- AMARH826636
- AMARH826638
- AMARH826640
- AMARH826642
- AMARH826644
- AMARH826646
- AMARH826648
- AMARH826786DKD3
- AMARH826788DKD3
- AMARH826790DKD3
- AMARH826640/DKD1
- AMARH826786WC
- AMARH826640/DKD3
- AMARH826788WC
- Accuracy:
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- Reference temperature:
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
Specifications
Product Family Options
With thermometer
Product Information
- ModelRangeDivisionTemperature RangeLengthAvailabilityPrice
- AMARH826760-FLSpecific gravity hydrometer, filling: with red or blue special liquidSpecific gravity hydrometer, filling: with red or blue special liquid101 H0,500 - 0,650 °SG0,001 °SG+30...+90 °F; –1,1...+32,2 °C360 mmLog in for availability1 * 1 ST194,00 €Specifications:
More
Specifications
Cat. No.AMARH826760-FLAccuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826764-FLSpecific gravity hydrometer, filling: with red or blue special liquidSpecific gravity hydrometer, filling: with red or blue special liquid310 H500 - 650 °SG0,001 °SG0...0,5 °C; 35...0,5 °C390 mmLog in for availability1 * 1 ST212,00 €Specifications:
More
Specifications
Cat. No.AMARH826764-FLAccuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C)
Without thermometer
Product Information
- ModelRangeDivisionTemperature RangeLengthAvailabilityPrice
- AMARH826780Specific gravity hydrometerSpecific gravity hydrometer102 H - 620,650 - 0,700 °SG0,001 °SG260 mmLog in for availability1 * 1 ST45,70 €Specifications:
More
Specifications
Cat. No.AMARH826780Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826782Specific gravity hydrometerSpecific gravity hydrometer103 H - 620,700 - 0,750 °SG0,001 °SG260 mmLog in for availability1 * 1 ST45,70 €Specifications:
More
Specifications
Cat. No.AMARH826782Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826784Specific gravity hydrometerSpecific gravity hydrometer104 H - 620,750 - 0,800 °SG0,001 °SG260 mmLog in for availability1 * 1 ST45,70 €Specifications:
More
Specifications
Cat. No.AMARH826784Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826786Specific gravity hydrometerSpecific gravity hydrometer105 H - 620,800 - 0,850 °SG0,001 °SG260 mmLog in for availability1 * 1 ST45,70 €Specifications:
More
Specifications
Cat. No.AMARH826786Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826788Specific gravity hydrometerSpecific gravity hydrometer106 H - 620,850 - 0,900 °SG0,001 °SG260 mmLog in for availability1 * 1 ST45,70 €Specifications:
More
Specifications
Cat. No.AMARH826788Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826790Specific gravity hydrometerSpecific gravity hydrometer107 H - 620,900 - 0,950 °SG0,001 °SG260 mmLog in for availability1 * 1 ST45,70 €Specifications:
More
Specifications
Cat. No.AMARH826790Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826792Specific gravity hydrometerSpecific gravity hydrometer108 H - 620,950 - 1,000 °SG0,001 °SG260 mmLog in for availability1 * 1 ST45,70 €Specifications:
More
Specifications
Cat. No.AMARH826792Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826910Specific gravity hydrometerSpecific gravity hydrometer125 H - 621,000 - 1,050 °SG0,001 °SG260 mmLog in for availability1 * 1 ST47,40 €Specifications:
More
Specifications
Cat. No.AMARH826910Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826912Specific gravity hydrometerSpecific gravity hydrometer126 H - 621,050 - 1,100 °SG0,001 °SG260 mmLog in for availability1 * 1 ST47,40 €Specifications:
More
Specifications
Cat. No.AMARH826912Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826914Specific gravity hydrometerSpecific gravity hydrometer127 H - 621,100 - 1,150 °SG0,001 °SG260 mmLog in for availability1 * 1 ST47,40 €Specifications:
More
Specifications
Cat. No.AMARH826914Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826916Specific gravity hydrometerSpecific gravity hydrometer128 H - 621,150 - 1,200 °SG0,001 °SG260 mmLog in for availability1 * 1 ST47,40 €Specifications:
More
Specifications
Cat. No.AMARH826916Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826918Specific gravity hydrometerSpecific gravity hydrometer129 H - 621,200 - 1,250 °SG0,001 °SG260 mmLog in for availability1 * 1 ST47,40 €Specifications:
More
Specifications
Cat. No.AMARH826918Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826920Specific gravity hydrometerSpecific gravity hydrometer130 H - 621,250 - 1,300 °SG0,001 °SG260 mmLog in for availability1 * 1 ST47,40 €Specifications:
More
Specifications
Cat. No.AMARH826920Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826922Specific gravity hydrometerSpecific gravity hydrometer131 H - 621,300 - 1,350 °SG0,001 °SG260 mmLog in for availability1 * 1 ST54,10 €Specifications:
More
Specifications
Cat. No.AMARH826922Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826924Specific gravity hydrometerSpecific gravity hydrometer132 H - 621,350 - 1,400 °SG0,001 °SG260 mmLog in for availability1 * 1 ST54,10 €Specifications:
More
Specifications
Cat. No.AMARH826924Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826926Specific gravity hydrometerSpecific gravity hydrometer133 H - 621,400 - 1,450 °SG0,001 °SG260 mmLog in for availability1 * 1 ST54,10 €Specifications:
More
Specifications
Cat. No.AMARH826926Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826928Specific gravity hydrometerSpecific gravity hydrometer134 H - 621,450 - 1,500 °SG0,001 °SG260 mmLog in for availability1 * 1 ST54,10 €Specifications:
More
Specifications
Cat. No.AMARH826928Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826930Specific gravity hydrometerSpecific gravity hydrometer135 H - 621,500 - 1,550 °SG0,001 °SG260 mmLog in for availability1 * 1 ST54,10 €Specifications:
More
Specifications
Cat. No.AMARH826930Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826932Specific gravity hydrometerSpecific gravity hydrometer136 H - 621,550 - 1,600 °SG0,001 °SG260 mmLog in for availability1 * 1 ST54,10 €Specifications:
More
Specifications
Cat. No.AMARH826932Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826934Specific gravity hydrometerSpecific gravity hydrometer137 H - 621,600 - 1,650 °SG0,001 °SG260 mmLog in for availability1 * 1 ST63,10 €Specifications:
More
Specifications
Cat. No.AMARH826934Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826936Specific gravity hydrometerSpecific gravity hydrometer138 H - 621,650 - 1,700 °SG0,001 °SG260 mmLog in for availability1 * 1 ST63,10 €Specifications:
More
Specifications
Cat. No.AMARH826936Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826938Specific gravity hydrometerSpecific gravity hydrometer139 H - 621,700 - 1,750 °SG0,001 °SG260 mmLog in for availability1 * 1 ST63,10 €Specifications:
More
Specifications
Cat. No.AMARH826938Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826940Specific gravity hydrometerSpecific gravity hydrometer140 H - 621,750 - 1,800 °SG0,001 °SG260 mmLog in for availability1 * 1 ST63,10 €Specifications:
More
Specifications
Cat. No.AMARH826940Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826942Specific gravity hydrometerSpecific gravity hydrometer141 H - 621,800 - 1,850 °SG0,001 °SG260 mmLog in for availability1 * 1 ST63,10 €Specifications:
More
Specifications
Cat. No.AMARH826942Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826500Specific gravity hydrometerSpecific gravity hydrometer82 H - 620,650 - 0,700 °SG0,005 °SG330 mmLog in for availability1 * 1 ST59,10 €Specifications:
More
Specifications
Cat. No.AMARH826500Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826502Specific gravity hydrometerSpecific gravity hydrometer83 H - 620,700 - 0,750 °SG0,005 °SG330 mmLog in for availability1 * 1 ST59,10 €Specifications:
More
Specifications
Cat. No.AMARH826502Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826504Specific gravity hydrometerSpecific gravity hydrometer84 H - 620,750 - 0,800 °SG0,005 °SG330 mmLog in for availability1 * 1 ST59,10 €Specifications:
More
Specifications
Cat. No.AMARH826504Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826506Specific gravity hydrometerSpecific gravity hydrometer85 H - 620,800 - 0,850 °SG0,005 °SG330 mmLog in for availability1 * 1 ST59,10 €Specifications:
More
Specifications
Cat. No.AMARH826506Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826508Specific gravity hydrometerSpecific gravity hydrometer86 H - 620,850 - 0,900 °SG0,005 °SG330 mmLog in for availability1 * 1 ST59,10 €Specifications:
More
Specifications
Cat. No.AMARH826508Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826510Specific gravity hydrometerSpecific gravity hydrometer87 H - 620,900 - 0,950 °SG0,005 °SG330 mmLog in for availability1 * 1 ST59,10 €Specifications:
More
Specifications
Cat. No.AMARH826510Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826512Specific gravity hydrometerSpecific gravity hydrometer88 H - 620,950 - 1,000 °SG0,005 °SG330 mmLog in for availability1 * 1 ST60,60 €Specifications:
More
Specifications
Cat. No.AMARH826512Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826628Specific gravity hydrometerSpecific gravity hydrometer98 H - 620,950 - 1,000 °SG0,005 °SG330 mmLog in for availability1 * 1 ST60,60 €Specifications:
More
Specifications
Cat. No.AMARH826628Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826514Specific gravity hydrometerSpecific gravity hydrometer89 H - 621,000 - 1,050 °SG0,005 °SG330 mmLog in for availability1 * 1 ST60,60 €Specifications:
More
Specifications
Cat. No.AMARH826514Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826630Specific gravity hydrometerSpecific gravity hydrometer111 H - 621,000 - 1,050 °SG0,005 °SG330 mmLog in for availability1 * 1 ST60,60 €Specifications:
More
Specifications
Cat. No.AMARH826630Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826516Specific gravity hydrometerSpecific gravity hydrometer90 H - 621,050 - 1,100 °SG0,005 °SG330 mmLog in for availability1 * 1 ST60,60 €Specifications:
More
Specifications
Cat. No.AMARH826516Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826632Specific gravity hydrometerSpecific gravity hydrometer112 H - 621,050 - 1,100 °SG0,005 °SG330 mmLog in for availability1 * 1 ST60,60 €Specifications:
More
Specifications
Cat. No.AMARH826632Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826634Specific gravity hydrometerSpecific gravity hydrometer113 H - 621,100 - 1,150 °SG0,005 °SG330 mmLog in for availability1 * 1 ST60,60 €Specifications:
More
Specifications
Cat. No.AMARH826634Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826636Specific gravity hydrometerSpecific gravity hydrometer114 H - 621,150 - 1,200 °SG0,005 °SG330 mmLog in for availability1 * 1 ST60,60 €Specifications:
More
Specifications
Cat. No.AMARH826636Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826638Specific gravity hydrometerSpecific gravity hydrometer115 H - 621,200 - 1,250 °SG0,005 °SG330 mmLog in for availability1 * 1 ST60,60 €Specifications:
More
Specifications
Cat. No.AMARH826638Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826640Specific gravity hydrometerSpecific gravity hydrometer116 H - 621,250 - 1,300 °SG0,005 °SG330 mmLog in for availability1 * 1 ST60,60 €Specifications:
More
Specifications
Cat. No.AMARH826640Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826642Specific gravity hydrometerSpecific gravity hydrometer117 H - 621,300 - 1,350 °SG0,005 °SG330 mmLog in for availability1 * 1 ST105,00 €Specifications:
More
Specifications
Cat. No.AMARH826642Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826644Specific gravity hydrometerSpecific gravity hydrometer118 H - 621,350 - 1,400 °SG0,005 °SG330 mmLog in for availability1 * 1 ST105,00 €Specifications:
More
Specifications
Cat. No.AMARH826644Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826646Specific gravity hydrometerSpecific gravity hydrometer119 H - 621,400 - 1,450 °SG0,005 °SG330 mmLog in for availability1 * 1 ST105,00 €Specifications:
More
Specifications
Cat. No.AMARH826646Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826648Specific gravity hydrometerSpecific gravity hydrometer120 H - 621,450 - 1,500 °SG0,005 °SG330 mmLog in for availability1 * 1 ST105,00 €Specifications:
More
Specifications
Cat. No.AMARH826648Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C)
With DKD certificate
Product Information
- ModelRangeDivisionTemperature RangeLengthAvailabilityPrice
- AMARH826786DKD3Specific gravity hydrometerSpecific gravity hydrometer105 H - 620,800 - 0,850 °SG0,001 °SG260 mmLog in for availability1 * 1 STSpecifications:
More
Specifications
Cat. No.AMARH826786DKD3Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826788DKD3Specific gravity hydrometerSpecific gravity hydrometer106 H - 620,850 - 0,900 °SG0,001 °SG260 mmLog in for availability1 * 1 STSpecifications:
More
Specifications
Cat. No.AMARH826788DKD3Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826790DKD3Specific gravity hydrometerSpecific gravity hydrometer107 H - 620,900 - 0,950 °SG0,001 °SG260 mmLog in for availability1 * 1 STSpecifications:
More
Specifications
Cat. No.AMARH826790DKD3Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826640/DKD1Specific gravity hydrometerSpecific gravity hydrometer116 H - 621,250 - 1,300 °SG0,005 °SG330 mmLog in for availability1 * 1 STSpecifications:
More
Specifications
Cat. No.AMARH826640/DKD1Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826640/DKD3Specific gravity hydrometerSpecific gravity hydrometer116 H - 621,250 - 1,300 °SG0,005 °SG330 mmLog in for availability1 * 1 STSpecifications:
More
Specifications
Cat. No.AMARH826640/DKD3Accuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C)
With factory test certificate
Product Information
- ModelRangeDivisionTemperature RangeLengthAvailabilityPrice
- AMARH826786WCSpecific gravity hydrometerSpecific gravity hydrometer105 H - 620,800 - 0,850 °SG0,001 °SG260 mmLog in for availability1 * 1 ST45,70 €Specifications:
More
Specifications
Cat. No.AMARH826786WCAccuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826788WCSpecific gravity hydrometerSpecific gravity hydrometer106 H - 620,850 - 0,900 °SG0,001 °SG260 mmLog in for availability1 * 1 ST45,70 €Specifications:
More
Specifications
Cat. No.AMARH826788WCAccuracy±1 scale divisionReference temperature60/60 °F (except model AMARH826764-FL: 15 °C)