VWR®, IOS W-Plan Objectives
Supplier: VWR Collection
Jump to:
About this item
These high contrast objectives are ideal for routine observations in laboratories.
- Ensure field flatness up to F.N. 22
- Based on infinity-corrected optical system
- 100× is also available with iris for darkfield
8 Options Available Below
Product Details & Documents
These high contrast objectives are ideal for routine observations in laboratories.
- Ensure field flatness up to F.N. 22
- Based on infinity-corrected optical system
- 100× is also available with iris for darkfield
Documents
VWR® for Microscopy
Product Literature
Product Family Options
Product Information
- MagnificationAvailabilityPrice
- 630-2948IOS W-Plan objective
- 630-2949IOS W-Plan objective
- 630-2950IOS W-Plan objective
- 630-2951IOS W-Plan objective
- 630-2952IOS W-Plan objective
- 630-2953IOS W-Plan objective
- 630-2955IOS W-Plan objective (oil)
- 630-2954IOS W-Plan objective (oil) with iris for DarkfieldIOS W-Plan objective (oil) with iris for Darkfield100×/1,25 (i)Log in for availability1 * 1 ST612,00 €