ContactUsLinkComponent
Media in bottles, sabouraud dextrose broth
Media in bottles, sabouraud dextrose broth
Catalog # LIOF402040
Supplier:  LIOFILCHEM
CAS Number:  
Media in bottles, sabouraud dextrose broth
Catalog # LIOF402040
Supplier:  LIOFILCHEM
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Sabouraud dextrose broth, 6×100 ml
  • Filling:
    100 ml
Specifications
About this item
Sabouraud dextrose broth (SDB) is a liquid medium used in qualitative procedures for isolation of yeasts and moulds and for the culture or subculture of fungi from clinical and nonclinical specimens.
Certifications: This medium conforms to the requirements of the harmonised method in the United States Pharmacopoeia (USP), European Pharmacopoeia (Ph. Eur.) and Japanese Pharmacopoeia (JP) for the microbiological examination of non sterile products.