Magnetic stirring bars, cylindrical, PTFE, fully traceable
Supplier: COWIE
Certificates
About this item
These cylindrical stirring bars feature a smooth, round profile and are ideal for a wide variety of applications.
- Manufactured from virgin PTFE materials with FDA and USP Class VI approvals
- With alnico magnet core
- Chemically inert, non-contaminating and usable to 280 °C
- Isostatic encapsulation to eliminate cracks and porosity
- Polished finish to reduce pick-up and cross contamination
Used for stirring and mixing in lquid media and in open and closed systems over a broad temperature range. These bars can be sterilised by chemical or thermal means, but not by gamma radiation.
Certifications: ISO 9001:2015.
Packaging: Packed in a grip seal bag.
Frequently Bought Together