ContactUsLinkComponent
Fluprostenol ELISA kit
Fluprostenol ELISA kit
Supplier:  Cayman Chemical
Certificates
undefined
Fluprostenol ELISA kit
Supplier:  Cayman Chemical
Specifications
  • Developmental time:
    90 - 120 min
  • Detection range:
    7,8 - 1000 pg/ml
  • Sensitivity:
    16 pg/ml
  • Incubation time:
    18 hr