Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Carbon Standards
Single element standards for ICP-MS, carbon
Single element standards for ICP-MS, carbon
Single element standards for ICP-MS, carbon
Catalog # REGEPC2A7
Supplier:
REAGECON
CAS Number:
Expand image
Single element standards for ICP-MS, carbon
Catalog # REGEPC2A7
Supplier:
REAGECON
CAS Number:
Specifications
Concentration:
1000 mg/ml
Matrix:
H₂O
Pk:
100 mL
Environmentally Preferable:
Element:
Carbon
Symbol:
C
Specifications
About this item
Product Details
Product Details