Orders LinkContactUsLinkComponent
Pipettes, P-2, Aquisel
Pipettes, P-2, Aquisel
Aquisel pipette, P-2, with rubber stopper
Catalog # SELE1780225
Supplier:   SELECTA
Pipettes, P-2, Aquisel
Catalog # SELE1780225
Supplier:  SELECTA
Pipettes, P-2, Aquisel
Pipettes, P-2, Aquisel
Aquisel pipette, P-2, with rubber stopper
Price
Inquire For Price
Log in for availability
1 * 800 ST
Product Details & Documents
Polystyrene pipettes with zero-closure system allows the pipette filling, preventing that the blood exceeds the zero point and also avoiding total or partial loss.
  • Pipette filling by pressure till the bottom of the tube
  • For erythrocyte sedimentation read E.S.R. Westergren method
  • Compatible with conventional tubes of 12×86
  • Automatic sealing makes it a disposable product
Emballage: Cardboard package of 200 units. Supplied in 800 units boxes.
Specifications
  • Description:
    Aquisel pipette, P-2, with rubber stopper