Order Entry
Belgium
ContactUsLinkComponent
Soybean oil, epoxidised
Soybean oil, epoxidised
Catalog # SPCMS1179-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Soybean oil, epoxidised
Catalog # SPCMS1179-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Specific gravity at 25 °C:
    0.988 - 0.998
  • Acid value:
    0.50
  • Colour (APHA):
    125
  • Iodine Value:
    2.00
  • Moisture:
    0.10%

Specification Test Results

About this item

 :
 :975.399 g/mol
CAS Number:8013-07-8
Formula:C₅₇H₉₈O₁₂
MDL Number:MFCD00163560