Order Entry
Belgium
ContactUsLinkComponent
Periodic table mix 1 for ICP, 33 elements, TraceCERT®, Supelco®
Periodic table mix 1 for ICP, 33 elements, TraceCERT®, Supelco®
Catalog # 92091-100ML
Supplier:  Merck
CAS Number:  
Periodic table mix 1 for ICP, 33 elements, TraceCERT®, Supelco®
Catalog # 92091-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    10% HNO₃
  • Pk:
    100 mL
  • Description:
    Periodic table mix 1

Specifications

About this item

Periodic table mix 1 for ICP used as a standard for the quantitative determination of mineral composition in Tenualosa ilisha using inductively coupled plasma atomic emission spectroscopy (ICP-AES).

It contains Al, As, Ba, Be, Bi, B, Ca, Cd, Cs, Cr, Co, Cu, Ga, In, Fe, Pb, Li, Mg, Mn, Ni, P, K, Rb, Se, Si, Ag, Na, Sr, S, Te, Tl, V and Zn in 10% nitric acid (contains HF traces).

Certifications: Certified in accordance with ISO/IEC 17025 and ISO 17034