Order Entry
Belgium
Orders LinkContactUsLinkComponent
Fructanase

Fructanase

10 mL
Catalog # MEGAE-FRMXLQ
Supplier:   MEGAZYME
Fructanase
Catalog # MEGAE-FRMXLQ
Supplier:  MEGAZYME
CAS Number:  
Fructanase
Fructanase

10 mL

CAS Number:  
Price
374,00 €
Log in for availability
1 * 10 mL
Product Details & Documents

This purified fructanase mixture is used for the determination of fructn (liquid), in research, biochemical enzyme assays and in vitro diagnostic analysis.

This mixture consists of exo-inulinase 2000 U/ml (on kestose, at 40 °C), endo-inulinase ∼100 U/ml (on fructan at 40 °C), α-galactosidase <0,08 U/ml (on polygalacturonic acid at 40 °C), β-Glucanase <0,04 U/ml (on β-glucan at 40 °C) and pectinase <0,07 U/ml (on pectin at 40 °C).

This enzyme is supplied in a stabilised solution containing 50% glycerol and 0.02% sodium azide.

Specifications

Specifications

  • Pk:
    10 mL
  • Cat. No.:
    MEGAE-FRMXLQ
  • Storage conditions:
    −20 °C
  • Enzyme name:
    Fructanase

Specifications