Order Entry
Belgium
ContactUsLinkComponent
 
Tetrasodium diphosphate decahydrate ≥99.0% (by acidimetry) for analysis
  APPC121710.1211
 :  
Tetrasodium diphosphate decahydrate ≥99.0% (by acidimetry) for analysis
  APPC121710.1211
 :  

 

  • Pk:
    1.000 g
  • Seq:
    3047

 

 

  • Minimum Assay (Acidimetry):
    99.0%
  • pH of 5% Solution:
    9.5 - 10.7
  • Insoluble Matter in H₂O:
    ≤0.01%
  • Chloride (Cl):
    ≤0.005%
  • Sulphate (SO₄):
    ≤0.005%
  • Carbonate:
    Passes Test
  • Nitrate (NO₃):
    ≤0.003%
  • As:
    ≤0.0003%
  • Ca:
    ≤001%
  • Cu:
    ≤0.0005%
  • Fe:
    ≤0.0005%
  • MG:
    ≤0.01%
  • Ni:
    ≤0.0005%
  • PB:
    ≤0.0005%

 

 

 :
 :446.06 g/mol
 :79 °C
 :1.82
 :Ambient
 :231-767-1
 :13472-36-1
 :Na₄P₂O₇·10H₂O
 :MFCD00149200