Order Entry
Belgium
ContactUsLinkComponent
Haematoxylin solution Gill I, Sigma-Aldrich®
Corrosive
Irritant
Health
Haematoxylin solution Gill I, Sigma-Aldrich®
Catalog # GHS132-1L
CAS Number:  
undefined
Corrosive
Irritant
Health
Haematoxylin solution Gill I, Sigma-Aldrich®
Catalog # GHS132-1L
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :302.28 g/mol
 :M-Clarity™ quality level = MQ500 (more info.)
 :GHS05
 :GHS07
 :GHS08
 :1
CAS Number:517-28-2
 :208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111