Prostaglandin D₂-MOX Express ELISA kit
Supplier: Cayman Chemical
Certificates
About this item
A competitive assay that permits the rapid measurement of PGD₂ from biological samples.
- Treatment of samples with MOX hydrochloride converts unstable PGD₂ to stable PGD₂-MOX, MOX derivatization reagent included with assay
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure PGD₂-MOX levels down to 16 pg/ml
- Rapid assay; get results in under four hours
PGD₂-MOX Express ELISA is a competitive assay that permits the rapid measurement of PGD₂ from biological samples, requiring only one-hour incubation and development times for each step. The methodology for the assay is the same as that employed in the PGD₂-MOX ELISA (Item No. 512011). The ELISA offers the convenience of a fast assay while still achieving a sensitivity of ~16 pg/ml and an assay range of 7,8 to 1000 pg/ml.
Delivery: Kit contains: Antiserum, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, Ellman's reagent, control, methoxylamine, sodium acetate, tracer and antiserum dye.
Specifications
- Developmental time:60 min
- Mid point:60 - 100 pg/ml (50% B/B₀)
- Detection range:7,8 - 1000 pg/ml
- Storage temperature:–20 °C
- Sensitivity:16 pg/ml (80% B/B₀)
- Incubation time:1 hr
Specifications
Frequently Bought Together