Orders LinkContactUsLinkComponent
Drum wrench
Drum wrench
Drum wrench, Brass
Catalog # 119-0238
Drum wrench
Catalog # 119-0238
Drum wrench
Drum wrench, Brass
Price
57,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Drum Wrench opens drum bung caps.
  • Rugged and non-sparking
  • Design of handle protects the hand
This corrosive-resistant tool fits many 50,8 or 19,05 mm bung styles commonly used on drums.
Specifications
  • Description:
    Drum wrench
  • Material:
    Brass