ContactUsLinkComponent
Gel filtration chromatography columns, standard, CHROMAFLEX®
Gel filtration chromatography columns, standard, CHROMAFLEX®
Supplier:  DWK Life Sciences
Certificates
undefined
Gel filtration chromatography columns, standard, CHROMAFLEX®
Supplier:  DWK Life Sciences
Specifications
  • Catalog No:
  • SCERSP420830-6010
  • SCERSP420830-6020
  • SCERSP420830-3020
  • SCERSP420830-1220
  • SCERSP420830-1020
  • SCERSP420830-1210
  • SCERSP420830-3000
  • SCERSP420830-1520
  • SCERSP420830-1000
  • Fittings:
  • 1/4" - 28 flangeless fittings
  • 1/4" - 28 flangeless fittings
  • 1/4" - 28 flangeless fittings
  • 1/4" - 28 flangeless fittings
  • 1/4" - 28 flangeless fittings
  • 1/4" - 28 flangeless fittings
  • 1/4" - 28 flangeless fittings
  • 1/4" - 28 flangeless fittings
  • 1/4" - 28 flangeless fittings
  • Max. pressure:
  • 3,5 bar
  • 3,5 bar
  • 3,5 bar
  • 3,5 bar
  • 3,5 bar
  • 3,5 bar
  • 3,5 bar
  • 3,5 bar
  • 3,5 bar
  • Temperature limit:
  • 0...50 °C
  • 0...50 °C
  • 0...50 °C
  • 0...50 °C
  • 0...50 °C
  • 0...50 °C
  • 0...50 °C
  • 0...50 °C
  • 0...50 °C