Epitope retrieval system, decloaking Chamber™ NxGen
Catalog # BCARDC-2012
Supplier: BIOCARE MEDICAL
Specifications
- No. of slides:72 (3x24)
- Description:Decloaking Chamber™ NxGen, 110 V
- Seq:01
- W×D×H:361×343×330 mm
- Weight:6,91 kg
- Pk:1
Specifications
About this item
The Decloaking Chamber NxGen is an excellent tool for heat-induced epitope retrieval (HIER). The proper use of heat and pressure in conjunction with the appropriate buffer solutions is of the utmost importance for consistent immunohistochemistry (IHC) staining.
- Very gentle on tissues
- Steam monitor strips for temperature verification
- Audible alert upon run completion
- Total slide capacity of 72 and only minutes of hands-on time per run: Walk-away capability similar to fully automated staining instruments
- Five discrete temperature settings ranging between 60 °C and 110 °C with user programmable times
- 110 °C antigen retrieval protocol can be completed from start to finish in under an hour
- Data can be transferred to a PC via USB output
The Decloaking Chamber NxGen is designed to optimise and standardise antibody staining procedures and has been engineered to pass strict laboratory safety and quality control requirements. Temperature, pressure and time can be monitored and recorded to produce consistent staining.