


Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
:67-56-1
:
:CH₃OH
:MFCD00004595
:32.04 g/mol
:64...65 °C
:–97.8 °C
:0.792 kg/l
:200-659-6
:1230
:3 (6.1),II
:GHS02
:GHS06
:GHS08
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Minimum Assay (G.C.):99.9%
- Identity:IR Passes Test
- Density 20/4:0.791 - 0.79
- APHA colour:10
- Acidity:0.0002 meq/g
- Alkalinity:0.0002 meq/g
- Insoluble Matter in H₂O:Passes Test
- Non-Volatile Matter:0.0005%
- Reducing Substance to KMnO₄ (as O):0.00025%
- Darkened Substances by H₂SO₄:Passes Test
- Carbonyl Compounds (as CH₃COCH₃):0.002%
- Acetaldehyde (CH₃CHO):0.001%
- Acetone (G.C.):0.001%
- Formaldehyde (HCHO):0.001%
- Ethanol (G.C.):0.005%
- 2-Propanol (G.C.):0.005%
- Water(H₂O):0.05%
- Metals by ICP, Ag:0.05 mg/kg (ppm)
- Metals by ICP, Al:0.5 mg/kg (ppm)
- Metals by ICP, As:0.05 mg/kg (ppm)
- Metals by ICP, Au:0.05 mg/kg (ppm)
- Metals by ICP, B:0.02 mg/kg (ppm)
- Metals by ICP, Ba:0.1 mg/kg (ppm)
- Metals by ICP, Be:0.02 mg/kg (ppm)
- Metals by ICP, Bi:0.05 mg/kg (ppm)
- Metals by ICP, Ca:0.1 mg/kg (ppm)
- Metals by ICP, Cd:0.05 mg/kg (ppm)
- Metals by ICP, Co:0.02 mg/kg (ppm)
- Metals by ICP, Cr:0.02 mg/kg (ppm)
- Metals by ICP, Cu:0.02 mg/kg (ppm)
- Metals by ICP, Fe:0.1 mg/kg (ppm)
- Metals by ICP, Ga:0.02 mg/kg (ppm)
- Metals by ICP, Ge:0.05 mg/kg (ppm)
- Metals by ICP, Hg:0.05 mg/kg (ppm)
- Metals by ICP, In:0.05 mg/kg (ppm)
- Metals by ICP, K:0.1 mg/kg (ppm)
- Metals by ICP, Li:0.05 mg/kg (ppm)
- Metals by ICP, Mg:0.05 mg/kg (ppm)
- Metals by ICP, Mn:0.02 mg/kg (ppm)
- Metals by ICP, Mo:0.02 mg/kg (ppm)
- Metals by ICP, Na:0.5 mg/kg (ppm)
- Metals by ICP, Ni:0.02 mg/kg (ppm)
- Metals by ICP, P:0.2 mg/kg (ppm)
- Metals by ICP, Pb:0.02 mg/kg (ppm)
- Metals by ICP, Pt:0.02 mg/kg (ppm)
- Metals by ICP, S:0.2 mg/kg (ppm)
- Metals by ICP, Sb:0.02 mg/kg (ppm)
- Metals by ICP, Si:0.2 mg/kg (ppm)
- Metals by ICP, Sn:0.1 mg/kg (ppm)
- Metals by ICP, Sr:0.2 mg/kg (ppm)
- Metals by ICP, Ti:0.02 mg/kg (ppm)
- Metals by ICP, Tl:0.02 mg/kg (ppm)
- Metals by ICP, V:0.02 mg/kg (ppm)
- Metals by ICP, Zn:0.1 mg/kg (ppm)
- Metals by ICP, Zr:0.02 mg/kg (ppm)
Frequently Bought Together