Orders LinkContactUsLinkComponent
Assistent® Stirring Spatula
Assistent® Stirring Spatula
Stirring spatula with crossbar, polystyrene, -, Clear
Catalog # HECH40734010
Assistent® Stirring Spatula
Catalog # HECH40734010
Assistent® Stirring Spatula
Assistent® Stirring Spatula
Stirring spatula with crossbar, polystyrene, -, Clear
Price
9,80 €
Log in for availability
1 * 100 ST
Product Details & Documents
Designed for several uses, such as mixing samples and solutions in test tubes, resolution of substrate, additions of small reagent volumes, or releasing blood clots from the side of test tube walls.
  • Stirring rod for a wide variety of laboratory applications
Made from polystyrene, crystal clear.
Specifications
  • Sterile:
    -
  • Colour:
    Clear
  • Description:
    Stirring spatula with crossbar, polystyrene