Draining rack
Drying rack, height 105 mm
Catalog # HECH41238010
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Drying rack made of Plexiglas®, for 12 microscope slides.
The microscope slides are placed vertically on replaceable blotting paper. Two packs of paper can be stored in the base of the drying rack.
: Supplied with two packs of paper.
Specifications
- Désignation:Drying rack, height 105 mm