Orders LinkContactUsLinkComponent
Volumetric overflow flasks, without stopper
Volumetric overflow flasks, without stopper
Volumetric overflow flasks, without stopper
Certificates
Volumetric overflow flasks, without stopper
7 Options Available Below
Product Details & Documents
These volumetric overflow flasks are made of borosilicate glass 3.3.
  • Calibrated to deliver (TD, Ex)
  • Cut and ground exactly at the volume-mark
  • Without stopper
Specially designed for water analysis.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • NEUB0372-10-0435
    Overflow flask
    Overflow flask
    43,5 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,40 €
    26,40 €
    1 * 1 ST
       26,40 €
  • NEUB0372-10-094
    Overflow flask
    Overflow flask
    94 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,30 €
    31,30 €
    1 * 1 ST
       31,30 €
  • NEUB0372-10-100
    Overflow flask, neck: 17×1,8 mm
    Overflow flask, neck: 17×1,8 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,30 €
    35,30 €
    1 * 1 ST
       35,30 €
  • NEUB0372-10-164
    Overflow flask
    Overflow flask
    164 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,20 €
    50,20 €
    1 * 1 ST
       50,20 €
  • NEUB0372-10-250
    Overflow flask, neck: 17×1,8 mm
    Overflow flask, neck: 17×1,8 mm
    250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,70 €
    52,70 €
    1 * 1 ST
       52,70 €
  • NEUB0372-10-365
    Overflow flask
    Overflow flask
    365 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 78,90 €
    78,90 €
    1 * 1 ST
       78,90 €
  • NEUB0372-10-432
    Overflow flask
    Overflow flask
    432 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,30 €
    63,30 €
    1 * 1 ST
       63,30 €