ContactUsLinkComponent
Semivolatile calibration kit #3 (with benzidine)
Semivolatile calibration kit #3 (with benzidine)
Catalog # REST31463
Supplier:  Restek
CAS Number:  
undefined
Semivolatile calibration kit #3 (with benzidine)
Catalog # REST31463
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Semivolatile calibration kit #3 (with benzidine)
Specifications
About this item
Contains 1 ml each of the following mixtures, SV calibration mix #1 (anilines), SV calibration mix #2 (phenols), SV calibration mix #3 (base neutrals), SV calibration mix #4 (base neutrals), SV calibration mix #5 (PAHs), SV calibration mix #7 (dichlorobenzenes) and 605 benzidines calibration mix (benzidine and 3,3'-dichlorobenzidine).
Contains: 31007: SV Calibration Mix #1; Benzyl alcohol (100-51-6); 4-Chloroaniline (106-47-8); 2-Nitroaniline (88-74-4); 3-Nitroaniline (99-09-2); 4-Nitroaniline (100-01-6);31008: SV Calibration Mix #2; Benzoic acid (65-85-0); 4-Chloro-3-methylphenol (59-50-7); 2-Chlorophenol (95-57-8); 2,4-Dichlorophenol (120-83-2); 2,4-Dimethylphenol (105-67-9); 4,6-Dinitro-2-methylphenol (Dinitro-o-cresol) (534-52-1); 2,4-Dinitrophenol (51-28-5); 2-Methylphenol (o-cresol) (95-48-7); 4-Methylphenol (p-cresol) (106-44-5); 2-Nitrophenol (88-75-5); 4-Nitrophenol (100-02-7); Pentachlorophenol (87-86-5); Phenol (108-95-2); 2,4,5-Trichlorophenol (95-95-4); 2,4,6-Trichlorophenol (88-06-2);31009: SV Calibration Mix #3; Bis(2-chloroethoxy)methane (111-91-1); Bis(2-chloroethyl)ether (111-44-4); Bis(2-ethylhexyl)phthalate (117-81-7); 4-Bromophenyl phenyl ether (101-55-3); Benzyl butyl phthalate (85-68-7); 2-Chloronaphthalene (91-58-7); 4-Chlorophenyl phenyl ether (7005-72-3); Dimethylphthalate (131-11-3); Di-n-butylphthalate (84-74-2); Di-n-octylphthalate (117-84-0); N-Nitrosodimethylamine (62-75-9); N-Nitroso-di-n-propylamine (621-64-7); N-Nitrosodiphenylamine (86-30-6); 2,2'-Oxybis(1-chloropropane) (108-60-1);31010: SV Calibration Mix #4; Carbazole (86-74-8); Dibenzofuran (132-64-9); Diethyl phthalate (84-66-2); 2,4-Dinitrotoluene (121-14-2); 2,6-Dinitrotoluene (606-20-2); Hexachlorobenzene (118-74-1); Hexachlorobutadiene (87-68-3); Hexachlorocyclopentadiene (77-47-4); Hexachloroethane (67-72-1); Isophorone (78-59-1); 2-Methylnaphthalene (91-57-6); Nitrobenzene (98-95-3); 1,2,4-Trichlorobenzene (120-82-1);31011: SV Calibration Mix #5/610 PAH Mix; Acenaphthene (83-32-9); Acenaphthylene (208-96-8); Anthracene (120-12-7); Benz(a)anthracene (56-55-3); Benzo(a)pyrene (50-32-8); Benzo(b)fluoranthene (205-99-2); Benzo(k)fluoranthene (207-08-9); Benzo(ghi)perylene (191-24-2); Chrysene (218-01-9); Dibenz(a,h)anthracene (53-70-3); Fluoranthene (206-44-0); Fluorene (86-73-7); Indeno(1,2,3-cd)pyrene (193-39-5); Naphthalene (91-20-3); Phenanthrene (85-01-8); Pyrene (129-00-0);31013: SV Calibration Mix #7; 1,2-Dichlorobenzene (95-50-1); 1,3-Dichlorobenzene (541-73-1); 1,4-Dichlorobenzene (106-46-7);31030: 605 Benzidines Calibration Mix; Benzidine (92-87-5); 3,3'-Dichlorobenzidine (91-94-1).
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.