Order Entry
Belgium
ContactUsLinkComponent
8270 calibration mix #5, revised (18 components)
8270 calibration mix #5, revised (18 components)
Catalog # REST31995
Supplier:  Restek
CAS Number:  
undefined
8270 calibration mix #5, revised (18 components)
Catalog # REST31995
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in methylene chloride
  • Pk:
    1 mL
  • Description:
    8270 calibration mix #5, revised
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Contains: Acenaphthene (83-32-9); Acenaphthylene (208-96-8); Anthracene (120-12-7); Benz(a)anthracene (56-55-3); Benzo(a)pyrene (50-32-8); Benzo(b)fluoranthene (205-99-2); Benzo(ghi)perylene (191-24-2); Benzo(k)fluoranthene (207-08-9); Chrysene (218-01-9); Dibenz(a,h)anthracene (53-70-3); Fluoranthene (206-44-0); Fluorene (86-73-7); Indeno(1,2,3-cd)pyrene (193-39-5); 1-Methylnaphthalene (90-12-0); 2-Methylnaphthalene (91-57-6); Naphthalene (91-20-3); Phenanthrene (85-01-8); Pyrene (129-00-0)

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.