Order Entry
Belgium
ContactUsLinkComponent
508.1 GC degradation check mix (2 components)
508.1 GC degradation check mix (2 components)
Catalog # REST32093
Supplier:  Restek
CAS Number:  
undefined
508.1 GC degradation check mix (2 components)
Catalog # REST32093
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    100 μg/ml each in ethyl acetate
  • Pk:
    1
  • Description:
    508.1 GC degradation check mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Conatains two components, those are 4,4'-DDT and endrin.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.