508.1 surrogate (4,4'-dibromobiphenyl)
Catalog # REST32092
Supplier: Restek
CAS Number:
Specifications
- Concentration:500 μg/ml in ethyl acetate
- Pk:1
- Description:508.1 surrogate (4,4'-dibromobiphenyl)
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.