Amine column test mix (GC) (8 components)
Catalog # REST35002
Supplier: Restek
CAS Number:
Specifications
- Pk:1 mL
- Description:Amine column test mix, in methylene chloride:methanol (1:1)
- Pk Info:1 ml/ampoule
Specifications
About this item
The amine column test mix consists of (C10) decane (0,60 mg/ml), (C12) dodecane (0.60 mg/ml, 1,2-butanediol (0.60 mg/ml), diethanolamine (DEA) (1.20 mg/ml), diethylenetriamine (1.20 mg/ml), 2,6-dimethylaniline (0.60 mg/ml), 2-nonanol (0.60 mg/ml), pyridine(0.60 mg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.