Order Entry
Belgium
ContactUsLinkComponent
GC-MS tuning mixture (4 components)
GC-MS tuning mixture (4 components)
Catalog # REST31615
Supplier:  Restek
CAS Number:  
undefined
GC-MS tuning mixture (4 components)
Catalog # REST31615
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    1000 μg/ml each in methylene chloride
  • Pk:
    1 mL
  • Description:
    GC-MS tuning mixture
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Contains four components, those are benzidine, 4,4'-DDT, decafluorotriphenylphosphine and pentachlorophenol.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.