Order Entry
Belgium
ContactUsLinkComponent
WA EPH fractionation check mix (22 components)
WA EPH fractionation check mix (22 components)
Catalog # REST31491
Supplier:  Restek
CAS Number:  
undefined
WA EPH fractionation check mix (22 components)
Catalog # REST31491
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    25 μg/ml each in hexane
  • Pk:
    1 mL
  • Description:
    WA EPH fractionation check mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The mix consists of, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, (C16) n-hexadecane, (C21) n-heneicosane, (C34) n-tetratriacontane, acenaphthene, acenaphthylene, anthracene, benz(a)anthracene, benzo(a)pyrene, benzo(b)fluoranthene, benzo(k)fluoranthene, benzo(ghi)perylene, chrysene, dibenz(a,h)anthracene, fluoranthene, fluorene, indeno(1,2,3-cd)pyrene, naphthalene, phenanthrene, pyrene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.