ContactUsLinkComponent
WA EPH aliphatic hydrocarbon mix (6 components)
WA EPH aliphatic hydrocarbon mix (6 components)
  REST31489
 :  Restek
 :  
undefined
WA EPH aliphatic hydrocarbon mix (6 components)
  REST31489
 :  Restek
 :  
  • Concentration:
    1000 μg/ml each in hexane
  • Pk:
    1
  • Description:
    WA EPH aliphatic hydrocarbon mix
  • Pk Info:
    1 ml/ampoule
The mix consists of, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, (C16) n-hexadecane, (C21) n-heneicosane, (C34) n-tetratriacontane.
 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.