WA EPH aromatic hydrocarbon mix (6 components)
Catalog # REST31488
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 μg/ml each in methylene chloride
- Pk:1
- Description:WA EPH aromatic hydrocarbon mix
- Pk Info:1 ml/ampoule
Specifications
About this item
The mix consists of, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, (C16) n-hexadecane, (C21) n-heneicosane, (C34) n-tetratriacontane.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.