Order Entry
Belgium
ContactUsLinkComponent
Alternate boiling point/carbon number distribution marker stock standard (9 components)
Alternate boiling point/carbon number distribution marker stock standard (9 components)
Catalog # REST31639
Supplier:  Restek
CAS Number:  
undefined
Alternate boiling point/carbon number distribution marker stock standard (9 components)
Catalog # REST31639
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    200 μg/ml each in pentane
  • Pk:
    1 mL
  • Description:
    Alternate boiling point/carbon number distribution marker stock standard
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The mix consists of (C6) n-hexane, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, (C16) n-hexadecane, (C21) n-heneicosane, (C28) n-octacosane, (C35) n-pentatriacontane, (C36) n-hexatriacontane.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.