TX TPH matrix spike mix (2 components)
REST31484
: Restek
:
- Concentration:10000 μg/ml each in P and T methanol
- Pk:1
- Description:TX TPH matrix spike mix
- Pk Info:1 ml/ampoule
The mix consists of diesel fuel #2 composite, unleaded gasoline composite.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.