Saisie de commande
Belgium
ContactUsLinkComponent
MA VPH matrix spike mix with surrogate rev. 1.1 (July 2004) (16 components)
MA VPH matrix spike mix with surrogate rev. 1.1 (July 2004) (16 components)
Catalog # REST30605
Supplier:  Restek
CAS Number:  
undefined
MA VPH matrix spike mix with surrogate rev. 1.1 (July 2004) (16 components)
Catalog # REST30605
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    50 μg/ml in P and T methanol
  • Cdt:
    1 Ampoul
  • Désignation:
    MA VPH matrix spike mix with surrogate rev. 1.1
  • Info cdt:
    1 ml/ampoule

Specifications

About this item

The mix consists of (C5) n-pentane, (C9) n-nonane, (C10) n-decane, benzene, n-butylcyclohexane, 2,5-dibromotoluene (SS), ethylbenzene, 2-methylpentane, methyl tert-butyl ether, naphthalene, toluene, 1,2,4-trimethylbenzene, 2,2,4-trimethylpentane, m-xylene, o-xylene, p-xylene.

  • For current Massachusetts VPH method - surrogate included
  • Matrix spike mix at 50 μg/ml, per updated method
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.