Certified PAHs in diesel
Catalog # REST31673
Supplier: Restek
CAS Number:
Specifications
- Concentration:50000 ppm diesel #2 in methylene chloride
- Pk:1
- Description:Certified PAHs in diesel
- Pk Info:1 ml/ampoule
Specifications
About this item
The mix consists of, acenaphthene, 2-methylnaphthalene, acenaphthylene, fluorene, naphthalene, 1-methylnaphthalene and phenanthrene.
- Confirms diesel #2 TPH and priority PAHs in a single analysis
- Complete data packs available
Certificate of analysis includes concentration of TPH and certified concentrations of individual PAHs.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.