515.4 calibration mix (16 components)
Catalog # REST32443
Supplier: Restek
CAS Number:
Specifications
- Pk:1 mL
- Description:515.4 calibration mix, in acetone
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains 16 components, those are acifluorfen, bentazon, chloramben, 2,4-d, dalapon, 2,4-db, dcpa diacid, dicamba, 3,5-dichlorobenzoic acid, dichlorprop, dinoseb, pentachlorophenol, picloram, quinclorac, 2,4,5-t and 2,4,5-tp.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.