Paraquat and diquat calibration mix (2 components)
Catalog # REST32437
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 μg/ml each in water
- Pk:1 mL
- Description:Paraquat and diquat calibration mix
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains two components, those are diquat dibromide and paraquat dichloride.