MA EPH aromatic hydrocarbon standard (17 components)
Catalog # REST31458
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml each in methylene chloride
- Pk:1 mL
- Description:MA EPH aromatic hydrocarbon standard
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains: Acenaphthene, acenaphthylene, anthracene, benz(a)anthracene, benzo(a)pyrene, benzo(b)fluoranthene, benzo(k)fluoranthene, benzo(ghi)perylene, chrysene, dibenz(a,h)anthracene, fluoranthene, fluorene, indeno(1,2,3-cd)pyrene, 2-methylnaphthalene, naphthalene, phenanthrene, pyrene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.